Tisk…

6-Hydroxybenzo[d]thiazole-2-carbonitrile 97%

Dodavatel: Apollo Scientific
Nebezpečí

APOSOR52126-1GEA 1250 CZK
APOSOR52126-1G
6-Hydroxybenzo[d]thiazole-2-carbonitrile 97%
6-Hydroxybenzo[d]thiazole-2-carbonitrile
Vzorec: C₈H₄N₂OS
MW: 176,19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00296905
CAS číslo: 939-69-5
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR