Diphenylacetaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
APOSOR59843-1GEA
1870
CZK
APOSOR59843-1G
APOSOR59843-5G
Diphenylacetaldehyde
Diphenylacetaldehyde
Vzorec:
C₁₄H₁₂O MW: 196,2400 g/mol Bod varu: 315 °C Hustota: 1,106 g/cm³ (25 °C) Bod vzplanutí: 113 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006972 CAS číslo: 947-91-1 EINECS: 213-433-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...