(S)-N-tert-Butoxycarbonylalanine methyl ester 95%
Dodavatel: Apollo Scientific
APOSOR61274-100GEA
3510
CZK
APOSOR61274-100G
(S)-N-tert-Butoxycarbonylalanine methyl ester 95%
(S)-N-tert-Butoxycarbonylalanine methyl ester
Vzorec:
C₉H₁₇NO₄ MW: 203,24 g/mol Bod tání: 32…35 °C Hustota: 1,03 g/cm³ (25 °C) Bod vzplanutí: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00038513 CAS číslo: 28875-17-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...