Tisk…

4-Chloro-1-methylpiperidine hydrochloride 97%

Dodavatel: Apollo Scientific
Varování

APOSOR6934-250GEA 9330 CZK
APOSOR6934-250G
4-Chloro-1-methylpiperidine hydrochloride 97%
4-Chloro-1-methylpiperidine hydrochloride
Vzorec: C₆H₁₃Cl₂N
MW: 170,08 g/mol
Bod tání: 158…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00012834
CAS číslo: 5382-23-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR