2,3,5-Trichloroisonicotinic acid
Dodavatel: Apollo Scientific
|
Varování
|
APOSOR70101-1GEA
2160
CZK
APOSOR70101-1G
2,3,5-Trichloroisonicotinic acid
2,3,5-Trichloroisonicotinic acid
Vzorec:
C₆H₂Cl₃NO₂ MW: 226,44 g/mol Bod tání: 193…194 °C Storage Temperature: Ambient |
MDL Number:
MFCD11044390 CAS číslo: 406676-18-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...