1-Methylimidazole-4-sulphonyl chloride 95+%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
APOSOR9010-25GEA
0
CZK
APOSOR9010-25G
1-Methylimidazole-4-sulphonyl chloride 95+%
1-Methylimidazole-4-sulphonyl chloride
Vzorec:
C₄H₅ClN₂O₂S MW: 180,61 g/mol Bod tání: 85…91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00068060 CAS číslo: 137049-00-4 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...