Tisk…

3-Phenoxypropiononitrile 95%

Dodavatel: Apollo Scientific

APOSOR911855-5GEA 3230 CZK
APOSOR911855-5G
3-Phenoxypropiononitrile 95%
3-Phenoxypropiononitrile
Vzorec: C₉H₉NO
Storage Temperature: Ambient
MDL Number: MFCD00013822
CAS číslo: 3055-86-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR