4'-Methoxy-2-chloroacetophenone 95%
Dodavatel: Apollo Scientific
APOSOR912451-1GEA
2370
CZK
APOSOR912451-1G
4'-Methoxy-2-chloroacetophenone 95%
4'-Methoxy-2-chloroacetophenone
Vzorec:
C₉H₉ClO₂ MW: 184,6196 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00216508 CAS číslo: 2196-99-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...