3-Hydroxydiphenylamine 95%
Dodavatel: Apollo Scientific
APOSOR920303-25GEA
5390
CZK
APOSOR920303-25G
APOSOR920303-100G
3-Hydroxydiphenylamine 95%
3-Hydroxydiphenylamine
Vzorec:
C₁₂H₁₁NO MW: 185,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00002262 CAS číslo: 101-18-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...