Tisk…

Ethyl-2,6-dimethylimidazo[1,2-a]pyridine-3-carboxylate

Dodavatel: Apollo Scientific

APOSOR9263-1GEA 6220 CZK
APOSOR9263-1G
Ethyl-2,6-dimethylimidazo[1,2-a]pyridine-3-carboxylate
Ethyl-2,6-dimethylimidazo[1,2-a]pyridine-3-carboxylate
Vzorec: C₁₂H₁₄N₂O₂
Storage Temperature: Ambient
CAS číslo: 81438-51-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR