2-Cyclopentylethanol 95%
Dodavatel: Apollo Scientific
APOSOR938017-5GEA
1910
CZK
APOSOR938017-5G
2-Cyclopentylethanol 95%
2-Cyclopentylethanol
Vzorec:
C₇H₁₄O MW: 114,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00019301 CAS číslo: 766-00-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...