3-Chloro-N,N-dimethylbenzamide 95%
Dodavatel: Apollo Scientific
APOSOR957133-1GEA
1830
CZK
APOSOR957133-1G
3-Chloro-N,N-dimethylbenzamide 95%
3-Chloro-N,N-dimethylbenzamide
Vzorec:
C₉H₁₀ClNO MW: 183,64 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01214035 CAS číslo: 24167-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...