Tisk…

2'-Fluoro-4'-nitroacetanilide

Dodavatel: Apollo Scientific

APOSPC300555-5GEA 4070 CZK
APOSPC300555-5G
2'-Fluoro-4'-nitroacetanilide
2'-Fluoro-4'-nitroacetanilide
Vzorec: C₈H₇FN₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00033912
CAS číslo: 348-19-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR