Tisk…

3-(3-Methyl-5-(trifluoromethoxy)phenyl)propionic acid 98%

Dodavatel: Apollo Scientific

APOSPC302510-1GEA 4860 CZK
APOSPC302510-1G
3-(3-Methyl-5-(trifluoromethoxy)phenyl)propionic acid 98%
3-(3-Methyl-5-(trifluoromethoxy)phenyl)propionic acid
Vzorec: C₁₁H₁₁F₃O₃
MW: 248,20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09025376
CAS číslo: 916420-57-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR