5-Chloro-2-(trifluoromethyl)benzylamine 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
APOSPC303095-1GEA
0
CZK
APOSPC303095-1G
5-Chloro-2-(trifluoromethyl)benzylamine 95%
5-Chloro-2-(trifluoromethyl)benzylamine
Vzorec:
C₈H₇ClF₃N MW: 209,60 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09832359 CAS číslo: 942319-05-3 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...