1,1,2,3,3,3-Hexafluoropropyl trichloromethyl ether
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:56860-83-4
:
Formula:C₄HCl₃F₆O
MDL Number:MFCD03094226
Storage Temperature:Ambient
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: PC3557-5GLocal Article Number: APOSPC3557-5G
- Supplier Article Number: PC3557-25GLocal Article Number: APOSPC3557-25G