Tisk…

7-Fluoro-2-methylquinoline-3-carboxylic acid

Dodavatel: Apollo Scientific

APOSPC404564-1GEA 10950 CZK
APOSPC404564-1G
7-Fluoro-2-methylquinoline-3-carboxylic acid
7-Fluoro-2-methylquinoline-3-carboxylic acid
Vzorec: C₁₁H₈FNO₂
Storage Temperature: Ambient
MDL Number: MFCD06254735
CAS číslo: 879361-44-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR