4-(Dimethylcarbamoyl)-2-fluorophenylboronic acid 95%
Dodavatel: Apollo Scientific
APOSPC412105-1GEA
14090
CZK
APOSPC412105-1G
4-(Dimethylcarbamoyl)-2-fluorophenylboronic acid 95%
4-(Dimethylcarbamoyl)-2-fluorophenylboronic acid
Vzorec:
C₉H₁₁BFNO₃ MW: 211,00 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11855833 CAS číslo: 874289-30-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...