Tisk…

2-Cyano-6-fluorophenylboronic acid 95+%

Dodavatel: Apollo Scientific

APOSPC412336-250MGEA 2330 CZK
APOSPC412336-250MG
2-Cyano-6-fluorophenylboronic acid 95+%
2-Cyano-6-fluorophenylboronic acid
Vzorec: C₇H₅BFNO₂
MW: 164,9295 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09033623
CAS číslo: 656235-44-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR