Tisk…

3-(3'-Fluorobenzyloxy)phenylboronic acid

Dodavatel: Apollo Scientific

APOSPC430505-250MGEA 2070 CZK
APOSPC430505-250MG
3-(3'-Fluorobenzyloxy)phenylboronic acid
3-(3'-Fluorobenzyloxy)phenylboronic acid
Vzorec: C₁₃H₁₂BFO₃
MW: 246,0420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08276822
CAS číslo: 1072951-62-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR