Tisk…

4-Fluoro-3-(methoxy(methyl)carbamoyl)phenylboronic acid 98%

Dodavatel: Apollo Scientific

APOSPC5093-1GEA 5000 CZK
APOSPC5093-1G
4-Fluoro-3-(methoxy(methyl)carbamoyl)phenylboronic acid 98%
4-Fluoro-3-(methoxy(methyl)carbamoyl)phenylboronic acid
Vzorec: C₉H₁₁BFNO₄
MW: 227,00 g/mol
Bod tání: 160…163 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08235106
CAS číslo: 874290-69-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR