Tisk…

6-(3-Fluorophenyl)-2-pyridinecarboxylic acid

Dodavatel: Apollo Scientific

APOSPC510349-1GEA 14060 CZK
APOSPC510349-1G
6-(3-Fluorophenyl)-2-pyridinecarboxylic acid
6-(3-Fluorophenyl)-2-pyridinecarboxylic acid
Vzorec: C₁₂H₈FNO₂
Storage Temperature: Ambient
MDL Number: MFCD06410053
CAS číslo: 887982-40-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR