4-Fluoro-3-methylbenzenesulphonyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
APOSPC5223-1GEA
4170
CZK
APOSPC5223-1G
4-Fluoro-3-methylbenzenesulphonyl chloride
4-Fluoro-3-methylbenzenesulphonyl chloride
Vzorec:
C₇H₆ClFO₂S MW: 208,63 g/mol Bod varu: 77…79 °C/0,1mmHg Bod tání: 35…40 °C Storage Temperature: Ambient |
MDL Number:
MFCD01365818 CAS číslo: 629672-19-1 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...