2-Bromo-5-fluorobenzamide
Dodavatel: Apollo Scientific
|
Varování
|
APOSPC5927-1GEA
1580
CZK
APOSPC5927-1G
2-Bromo-5-fluorobenzamide
2-Bromo-5-fluorobenzamide
Vzorec:
C₇H₅BrFNO Storage Temperature: Ambient |
MDL Number:
MFCD03094322 CAS číslo: 1006-34-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...