3-Bromo-4-fluorobiphenyl 97%
Dodavatel: Apollo Scientific
|
Varování
|
APOSPC6677-1GEA
830
CZK
APOSPC6677-1G
3-Bromo-4-fluorobiphenyl 97%
3-Bromo-4-fluorobiphenyl
Vzorec:
C₁₂H₈BrF MW: 251,10 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01571091 CAS číslo: 306935-88-6 UN: 3151 ADR: 9,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...