2-Fluoro-4'-chloroacetophenone 95%
Dodavatel: Apollo Scientific
APOSPC907972-1GEA
8100
CZK
APOSPC907972-1G
2-Fluoro-4'-chloroacetophenone 95%
2-Fluoro-4'-chloroacetophenone
Vzorec:
C₈H₆ClFO MW: 172,5840 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03412196 CAS číslo: 329-78-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...