3-Fluoro-5-iodobenzonitrile 98%
Dodavatel: Apollo Scientific
APOSPC9104-1GEA
3150
CZK
APOSPC9104-1G
3-Fluoro-5-iodobenzonitrile 98%
3-Fluoro-5-iodobenzonitrile
Vzorec:
C₇H₃FIN MW: 247,0100 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04116468 CAS číslo: 723294-75-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...