Tisk…

5-(2-Fluorophenyl)isoxazole

Dodavatel: Apollo Scientific

Varování

APOSPC9960-250MGEA 1040 CZK
APOSPC9960-250MG
5-(2-Fluorophenyl)isoxazole
5-(2-Fluorophenyl)isoxazole
Vzorec: C₉H₆FNO
MW: 163,15 g/mol
Bod varu: 68…70 °C/0,3mmHg
Storage Temperature: Ambient
MDL Number: MFCD02183542
CAS číslo: 138716-36-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR