Tisk…
Yeast alpha-glucosidase
Dodavatel: Apollo Scientific
APOSBIB3066-100KUEA 18650 CZK
APOSBIB3066-100KU
Yeast alpha-glucosidase
Enzymy
From yeast (Saccharomyces cerevisiae). Specific activity: IU/mg protein>50 units. IU per 1mg powder is approximately 70 units.