Tisk…
Sodium, plasma standard, BAKER INSTRA-ANALYZED® Reagent
BAKR5746.0100EA 4480 CZK
BAKR5746.0100
Sodium, plasma standard, BAKER INSTRA-ANALYZED® Reagent
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022