Order Entry
Nitsch and Nitsch basal salt mixture
Nitsch and Nitsch basal salt mixture
Supplier:  Apollo Scientific
Certificates
Nitsch and Nitsch basal salt mixture
Supplier:  Apollo Scientific
Product Information
  • Catalog #
    Supplier #
    Pk
    Availability
    Price
  • Nitsch and Nitsch basal salt mixture 10 L
    Nitsch and Nitsch basal salt mixture 10 L
    APOSPMN613-10L
    PMN613-10L
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L
  • Nitsch and Nitsch basal salt mixture 50 L
    Nitsch and Nitsch basal salt mixture 50 L
    APOSPMN613-50L
    PMN613-50L
    50 L
    Log in for availability
    Log in for availability
    1 * 50 L
    1 * 50 L