Order Entry
Multiple distributors with stopcock for bottles with GL 45 thread, BOLA
Multiple distributors with stopcock for bottles with GL 45 thread, BOLA
Supplier:  Bohlender
Certificates
undefined
Multiple distributors with stopcock for bottles with GL 45 thread, BOLA
Supplier:  Bohlender
Product Information
  • Catalog #
    Supplier #
    Thread
    Tube ext.Ø
    Neck
    Pk
    Availability
    Price
  • Two stopcocks with 4 mm hole
    Two stopcocks with 4 mm hole
    201-0298
    D616-08
    GL 45
    8 mm
    2 × GL 14
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 4 140,00 Kč
    4 140,00 Kč
    1 * 1 KS
       4 140,00 Kč
    Specifications:
    Specifications
    Environmentally Preferable
  • Three stopcocks with 4 mm hole
    Three stopcocks with 4 mm hole
    201-0297
    D616-16
    GL 45
    8 mm
    3 × GL 14
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 4 280,00 Kč
    4 280,00 Kč
    1 * 1 KS
       4 280,00 Kč
    Specifications:
    Specifications
    Environmentally Preferable