Tisk…
Hledali jste: Gely elektroforetické
51  výsledků nalezeno
SearchResultCount:"51"
Sort Results
Zobrazit seznam Rychlý náhled (novinka)
Ohodnoťte výsledky hledání
Dodavatel: VWR Chemicals
Popis: An extensive line of acrylamide and bis-acrylamide pre-weighed powder blends, pre-mixed stock solutions and ready to use solutions for customised poly...
Dodavatel: VWR Chemicals
Popis: High quality standard agarose for routine analysis with broad separation range (0,05 to 50 kbp). Ideal for blotting applications due to high gel stren...
Dodavatel: VWR Chemicals
Popis: Agarose RA™ is a general application agarose that provides good resolution and is cost effective for high volume users. It was developed specifically ...
Dodavatel: VWR Chemicals
Popis: Agarose I™ is a standard melting/gelling agarose, suitable for routine nucleic acid analytical/preparative applications (250 bp to 22 kb).
Katalogové číslo: (E344-500ML)
Dodavatel: VWR Chemicals
Popis: Electrophoresis Gels, Acryl/Bis™, Acrylamide-Bis (29:1), 30% aqueous solution
Měrná jednotka: 1 * 500 mL
Katalogové číslo: (E347-500ML)
Dodavatel: VWR Chemicals
Popis: Electrophoresis Gels, Acryl/Bis™, Acrylamide-​Bis solution (37,5:1) 30% aqueous solution
Měrná jednotka: 1 * 500 mL
Katalogové číslo: (M157-500ML)
Dodavatel: VWR Chemicals
Popis: A 40% (w/v) solution of ultra pure acrylamide (38,96%) and bis-acrylamide (1,04%) at a final ratio of 37.5:1.
Měrná jednotka: 1 * 500 mL
Dodavatel: Biotium
Popis: High performance agarose for nucleic acid electrophoresis (analytical or preparative) and blotting. Validated for use with GelRed®, GelGreen®, and DNA...
Katalogové číslo: (PIER17856)
Dodavatel: Thermo Fisher Scientific
Popis: Agarose II (low melt) is a highly purified and carefully blended formulation that provides uniform lot-to-lot consistency for preparation of electroph...
Měrná jednotka: 1 * 100 g
Dodavatel: LONZA
Popis: SeaKem® LE is a high gel strength, low EEO, standard gelling temperature agarose perfect for electrophoresis and other molecular biology applications.
Dodavatel: LONZA
Popis: Latitude™ DNA gels are designed for accurate DNA fragment separation for almost any gel electrophoresis application. These ready-to-run gels are avail...
Dodavatel: LONZA
Popis: Reliant™ precast agarose minigels are designed for rapid and reproducible resolution of DNA fragments from 8 bp to 10 kb. Each precast agarose gel is ...
Dodavatel: EDVOTEK EUROPE LTD
Popis: 0.8% UltraSpec-Agarose™ prepared with TAE buffer. Simply melt, cool and pour.
Dodavatel: Thermo Fisher Scientific
Popis: Agarose I (molecular biology grade) is a highly purified and carefully blended formulation that provides uniform lot-to-lot consistency for preparatio...
Dodavatel: LONZA
Popis: Intermediate melting point agarose ideal for resolution of DNA fragments, PCR and RT-PCR products, differing in size by 2%, in the range of 20 to 800 ...
Katalogové číslo: (LONZ50302)
Dodavatel: LONZA
Popis: Ultra-soft agarose, ideal for high efficiency hybridoma cloning and expansion of cDNA libraries.
Měrná jednotka: 1 * 25 g
Cena na vyžádání
Omezené množství produktu na skladě. Zboží může být k dispozici v jiném skladě poblíž vašeho sídla. Přesvědčte se, že jste přihlášení na stránky, abyste mohli vidět dostupné položky na skladě. Pokud je stále zobrazeno call a potřebujete asistenci, volejte na číslo 321 570 321.
Omezené množství produktu na skladě. Zboží může být k dispozici v jiném skladě poblíž vašeho sídla. Přesvědčte se, že jste přihlášení na stránky, abyste mohli vidět dostupné položky na skladě. Pokud je stále zobrazeno call a potřebujete asistenci, volejte na číslo 321 570 321.
Daná chemikálie je regulována dle platné legislativy a bude vyžadováno vyplnění formuláře. Jeho včasným vyplněním uspíšíte dodání produktu.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Tento produkt je zablokován vaší organizací. Kontaktujte své nákupní oddělení pro více informací.
Původní produkt již není k dispozici. Zobrazen je náhradní produkt.
Tento produkt již není k dispozici. Podobné produkty můžete vyhledat pomocí VWR katalogových čísel nebo čísel dodavatele uvedených výše. Pokud potřebujete další asistenci, zavolejte na telefonní číslo zákaznického servisu VWR 321 570 321.
1 - 16 of 51
no targeter for Bottom