Tisk…
Hledali jste: Pojidla
15  výsledků nalezeno
SearchResultCount:"15"
Sort Results
Zobrazit seznam Rychlý náhled (novinka)
Ohodnoťte výsledky hledání
Dodavatel: Thermo Scientific
Popis: High temperature adhesive for bonding ceramic to metal.
Katalogové číslo: (MMMAD400)
Dodavatel: 3M
Popis: 3M™ Scotch-Weld™ Structural adhesive nozzles are for use with 3M EPX™ Duo-Pak adhesives to ensure correct mixing of adhesive parts. Nozzle is matched ...
Měrná jednotka: 1 * 36 KS
Katalogové číslo: (293-0005)
Dodavatel: 3M
Popis: This reclosable fastener has a rubber based adhesive on the back of a black, polypropylene reclosable fastener. It's suitable for bonding many plastic...
Měrná jednotka: 1 * 1 Roll
Katalogové číslo: (MMMA90)
Dodavatel: 3M
Popis: 3M™ hi-strength 90 spray adhesive is an extremely versatile, fast-drying spray adhesive that bonds strongly to a wide range of materials. A permanent ...
Měrná jednotka: 1 * 12 KS
Katalogové číslo: (MMMA3764TCQ)
Dodavatel: 3M
Popis: The 3M™ Hot Melt Adhesive 3764 is a clear, 100% solids thermoplastic adhesive capable of bonding to many plastics, including polyolefin. Formulated to...
Měrná jednotka: 1 * 1 Roll
Katalogové číslo: (MMMA77)
Dodavatel: 3M
Popis: This is an extremely versatile, fast-drying, and transparent spray adhesive that bonds a wide range of lightweight materials. The permanent bond provi...
Měrná jednotka: 1 * 500 mL
Katalogové číslo: (819-0122)
Dodavatel: 3M
Popis: 3M™ repositionable spray adhesive 75 is a versatile, pressure sensitive, adhesive spray suitable for temporary holding or positioning of lightweight m...
Měrná jednotka: 1 * 12 KS
Katalogové číslo: (293-0009)
Dodavatel: 3M
Popis: This reclosable fastener is the convenient alternative to traditional fastening methods, such as screws, nuts, or bolts. It is designed to deliver a d...
Měrná jednotka: 1 * 1 Roll
Katalogové číslo: (293-0004)
Dodavatel: 3M
Popis: This high-performance PSA nylon hook and loop with a rubber adhesive and polypropylene film liner. It provides a firm adhesive bond to a wide variety ...
Měrná jednotka: 1 * 1 Roll
Katalogové číslo: (MMMAEPX400H)
Dodavatel: 3M
Popis: Manual applicator for dispensing EPX adhesive in 38 and 50 ml cartridges supplied with two plungers for 1:1/2:1 and 10:1 adhesive.
Měrná jednotka: 1 * 1 KS
Katalogové číslo: (293-0008)
Dodavatel: 3M
Popis: This reclosable fastener has a clear acrylic adhesive on the back of a clear polypropylene reclosable fastener with 40 stems/cm² and 5,8 mm engaged th...
Měrná jednotka: 1 * 1 Roll
Katalogové číslo: (MMMA190.50)
Dodavatel: 3M
Popis: Scotch-Weld™ Epoxy adhesive DP190 is a two-part epoxy adhesive that delivers exceptional performance with high shear and peel strength. Extremely comp...
Měrná jednotka: 1 * 12 KS
Katalogové číslo: (MMMA110T.400)
Dodavatel: 3M
Popis: Epoxy adhesive 110 Scotch-Weld™ 3M™ is a general purpose, fast setting, flexible, two-part epoxy adhesive.
Měrná jednotka: 1 * 400 mL
Katalogové číslo: (DOWC25100525)
Dodavatel: DOW CORNING
Popis: A grease-like silicone material, thickened with heat conductive metal oxides.
Měrná jednotka: 1 * 100 g
Katalogové číslo: (ICNA11FIXO0125)
Dodavatel: MP Biomedicals
Popis: Fixogum Rubber Cement is made for use in the formation of an air tight seal around the perimeter of the glass coverslip during probe hybridisation for...
Měrná jednotka: 1 * 125 mL
Cena na vyžádání
Omezené množství produktu na skladě. Zboží může být k dispozici v jiném skladě poblíž vašeho sídla. Přesvědčte se, že jste přihlášení na stránky, abyste mohli vidět dostupné položky na skladě. Pokud je stále zobrazeno call a potřebujete asistenci, volejte na číslo 321 570 321.
Omezené množství produktu na skladě. Zboží může být k dispozici v jiném skladě poblíž vašeho sídla. Přesvědčte se, že jste přihlášení na stránky, abyste mohli vidět dostupné položky na skladě. Pokud je stále zobrazeno call a potřebujete asistenci, volejte na číslo 321 570 321.
Daná chemikálie je regulována dle platné legislativy a bude vyžadováno vyplnění formuláře. Jeho včasným vyplněním uspíšíte dodání produktu.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Tento produkt je zablokován vaší organizací. Kontaktujte své nákupní oddělení pro více informací.
Původní produkt již není k dispozici. Zobrazen je náhradní produkt.
Tento produkt již není k dispozici. Podobné produkty můžete vyhledat pomocí VWR katalogových čísel nebo čísel dodavatele uvedených výše. Pokud potřebujete další asistenci, zavolejte na telefonní číslo zákaznického servisu VWR 321 570 321.
1 - 15 of 15
no targeter for Bottom