About this item
Whatman Puradisc syringe filters from Cytiva's business combine premium quality with economic efficiency. They are well suited for rapid, routine syringe filtration of samples up to 100 ml.
- Eight different material options
- Choice of 4 mm, 13 mm, and 30 mm syringe filter diameter to suit sample volume
- Range of pore sizes for retaining a variety of particles
- Optional sterile, medical-grade blister pack
- Optional tube-tip format for accurate dispensing into a micro-vial
- Recyclable after use
Puradisc syringe filters cater to a wide variety of sample types and applications, including capillary electrophoresis, biological preparations, protein filtration, and UV spectroscopy sample preparation. Different Puradisc membrane media support diverse chemical solvents and compounds.
Puradisc is available in the following membrane materials: regenerated cellulose (RC), cellulose nitrate, nylon, polytetrafluoroethylene (PTFE), polyethersulfone (PES), polypropylene, polyvinylidene difluoride (PVDF), and cellulose acetate.
Cytiva's Whatman Puradisc line is a comprehensive syringe filter portfolio used in a variety of applications. Cytiva also offers high-performance GD/X syringe filters with a multi-layer glass fiber prefilter for filtering larger volumes of harder-to-filter samples.
PVDF membranes are designed for high tensile strength, high solvent resistance, and low protein binding, making them suitable for biomedical filtration, sterilization filtration, and HPLC sample preparation.
Specifications
- Catalog No:
- 516-1124
- 516-1211
- 516-1213
- 516-1215
- 514-0223
- 514-0505
- 516-1111
- 514-0508
- 514-0506
- 514-0507
- 514-0499
- 516-0918
- 516-0919
- 514-0224
- 514-0225
- 514-0226
- 514-0227
- Ø:
- 13 mm
- 13 mm
- 13 mm
- 13 mm
- 25 mm
- 4 mm
- 4 mm
- 13 mm
- 4 mm
- 13 mm
- 13 mm
- 13 mm
- 13 mm
- 25 mm
- 25 mm
- 25 mm
- 25 mm
- Dimensions:
- 16,3×19,8 mm
- 16,3×19,8 mm
- 16,3×19,8 mm
- 16,3×19,8 mm
- 22,9×28,4 mm
- 10,1×23,5 mm
- 10,1×23,5 mm
- 16,3×19,8 mm
- 10,1×23,5 mm
- 16,3×19,8 mm
- 16,3×19,8 mm
- 16,3×19,8 mm
- 16,3×19,8 mm
- 22,9×28,4 mm
- 22,9×28,4 mm
- 22,9×28,4 mm
- 22,9×28,4 mm
- Max. operating pressure:
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- Weight:
- 0,95 g
- 0,95 g
- 0,95 g
- 0,95 g
- 2,7 g
- 0,55 g
- 0,55 g
- 0,95 g
- 0,55 g
- 0,95 g
- 0,95 g
- 0,95 g
- 0,95 g
- 2,7 g
- 2,7 g
- 2,7 g
- 2,7 g
Specifications
Product Information
- Catalog #Supplier #TypeØPore sizeVolumeMembraneInlet/outlet connectionsAvailabilityPrice
- 516-11116791-0402Puradisc™ 44 mm0,20 µm<2 mlPVDFFemale Luer lock/male LuerLog in for availability1 * 50 KS5 640,00 KčSupplier Article Number: 6791-0402Local Article Number: WHAT6791-0402Specifications:
More
Specifications
Environmentally PreferableØ4 mmDimensions10,1×23,5 mmMax. operating pressure5,2 barWeight0,55 g - 514-05056779-0402Puradisc™ 44 mm0,20 µm<2 mlPVDFFemale Luer lock/male LuerLog in for availability1 * 100 KS8 850,00 KčSupplier Article Number: 6779-0402Local Article Number: WHAT6779-0402Specifications:
More
Specifications
Environmentally PreferableØ4 mmDimensions10,1×23,5 mmMax. operating pressure5,2 barWeight0,55 g - 514-05066779-0404Puradisc™ 44 mm0,45 µm<2 mlPVDFFemale Luer lock/male LuerLog in for availability1 * 100 KS8 850,00 KčSupplier Article Number: 6779-0404Local Article Number: WHAT6779-0404Specifications:
More
Specifications
Environmentally PreferableØ4 mmDimensions10,1×23,5 mmMax. operating pressure5,2 barWeight0,55 g - 514-04996777-1302Puradisc™ 13 with tube tip13 mm0,20 µm<10 mlPVDFFemale Luer lock/male LuerLog in for availability1 * 50 KS5 990,00 KčSupplier Article Number: 6777-1302Local Article Number: WHAT6777-1302Specifications:
More
Specifications
Environmentally PreferableØ13 mmDimensions16,3×19,8 mmMax. operating pressure5,2 barWeight0,95 g - 516-12116791-1302Puradisc™ 1313 mm0,20 µm<10 mlPVDFFemale Luer lock/male LuerLog in for availability1 * 50 KS7 150,00 KčSupplier Article Number: 6791-1302Local Article Number: WHAT6791-1302Specifications:
More
Specifications
Environmentally PreferableØ13 mmDimensions16,3×19,8 mmMax. operating pressure5,2 barWeight0,95 g - 514-05076779-1302Puradisc™ 1313 mm0,20 µm<10 mlPVDFFemale Luer lock/male LuerLog in for availability1 * 100 KS8 600,00 KčSupplier Article Number: 6779-1302Local Article Number: WHAT6779-1302Specifications:
More
Specifications
Environmentally PreferableØ13 mmDimensions16,3×19,8 mmMax. operating pressure5,2 barWeight0,95 g - 516-12136792-1302Puradisc™ 1313 mm0,20 µm<10 mlPVDFFemale Luer lock/male LuerLog in for availability1 * 500 KS30 990,00 KčSupplier Article Number: 6792-1302Local Article Number: WHAT6792-1302Specifications:
More
Specifications
Environmentally PreferableØ13 mmDimensions16,3×19,8 mmMax. operating pressure5,2 barWeight0,95 g - 516-09186765-1302Puradisc™ 1313 mm0,20 µm<10 mlPVDFFemale Luer lock/male LuerLog in for availability1 * 2.000 KS87 910,00 KčSupplier Article Number: 6765-1302Local Article Number: WHAT6765-1302Specifications:
More
Specifications
Environmentally PreferableØ13 mmDimensions16,3×19,8 mmMax. operating pressure5,2 barWeight0,95 g - 514-05086779-1304Puradisc™ 1313 mm0,45 µm<10 mlPVDFFemale Luer lock/male LuerLog in for availability1 * 100 KS8 600,00 KčSupplier Article Number: 6779-1304Local Article Number: WHAT6779-1304Specifications:
More
Specifications
Environmentally PreferableØ13 mmDimensions16,3×19,8 mmMax. operating pressure5,2 barWeight0,95 g - 516-12156792-1304Puradisc™ 1313 mm0,45 µm<10 mlPVDFFemale Luer lock/male LuerLog in for availability1 * 500 KS30 770,00 KčSupplier Article Number: 6792-1304Local Article Number: WHAT6792-1304Specifications:
More
Specifications
Environmentally PreferableØ13 mmDimensions16,3×19,8 mmMax. operating pressure5,2 barWeight0,95 g - 516-09196765-1304Puradisc™ 1313 mm0,45 µm<10 mlPVDFFemale Luer lock/male LuerLog in for availability1 * 2.000 KS87 910,00 KčSupplier Article Number: 6765-1304Local Article Number: WHAT6765-1304Specifications:
More
Specifications
Environmentally PreferableØ13 mmDimensions16,3×19,8 mmMax. operating pressure5,2 barWeight0,95 g - 516-11246791-1304Puradisc™ 1313 mm0,45 µm<10 mlPVDFFemale Luer lock/male LuerLog in for availability1 * 50 KS6 310,00 KčSupplier Article Number: 6791-1304Local Article Number: WHAT6791-1304Specifications:
More
Specifications
Environmentally PreferableØ13 mmDimensions16,3×19,8 mmMax. operating pressure5,2 barWeight0,95 g - 514-02236746-2502Puradisc™ 2525 mm0,20 µm<100 mlPVDFFemale Luer lock/male LuerLog in for availability1 * 50 KS6 090,00 KčSupplier Article Number: 6746-2502Local Article Number: WHAT6746-2502Specifications:
More
Specifications
Environmentally PreferableØ25 mmDimensions22,9×28,4 mmMax. operating pressure5,2 barWeight2,7 g - 514-02256747-2502Puradisc™ 2525 mm0,20 µm<100 mlPVDFFemale Luer lock/male LuerLog in for availability1 * 200 KS19 700,00 KčSupplier Article Number: 6747-2502Local Article Number: WHAT6747-2502Specifications:
More
Specifications
Environmentally PreferableØ25 mmDimensions22,9×28,4 mmMax. operating pressure5,2 barWeight2,7 g - 514-02246746-2504Puradisc™ 2525 mm0,45 µm<100 mlPVDFFemale Luer lock/male LuerLog in for availability1 * 50 KS6 020,00 KčSupplier Article Number: 6746-2504Local Article Number: WHAT6746-2504Specifications:
More
Specifications
Environmentally PreferableØ25 mmDimensions22,9×28,4 mmMax. operating pressure5,2 barWeight2,7 g - 514-02266747-2504Puradisc™ 2525 mm0,45 µm<100 mlPVDFFemale Luer lock/male LuerLog in for availability1 * 200 KS19 700,00 KčSupplier Article Number: 6747-2504Local Article Number: WHAT6747-2504Specifications:
More
Specifications
Environmentally PreferableØ25 mmDimensions22,9×28,4 mmMax. operating pressure5,2 barWeight2,7 g - 514-02276749-2504Puradisc™ 2525 mm0,45 µm<100 mlPVDFFemale Luer lock/male LuerLog in for availability1 * 1.000 KS63 200,00 KčSupplier Article Number: 6749-2504Local Article Number: WHAT6749-2504Specifications:
More
Specifications
Environmentally PreferableØ25 mmDimensions22,9×28,4 mmMax. operating pressure5,2 barWeight2,7 g