Tisk…
MultiDrive basic M 20 successor package
Dodavatel: IKA
IKAA10005893EA 114100 CZK
IKAA10005893
MultiDrive basic M 20 successor package
Regardless of whether samples are hard, soft or fibrous – IKA MultiDrive basic performs a wide range of crushing tasks. In combination with the milling chamber MI 250 MultiDrive basic is perfectly suitable for dry grinding of hard and brittle substances.
  • High performance: 1000 Watt output and variable rotational speeds ranging from 3000 rpm to 20 000 rpm guarantee excellent crushing results.
  • Interval operation: Is an asset during the coarse crushing of hard samples or for extra thorough blending.
  • Integrated cooling: A cooling system is integrated in the milling cup, which allows indirect heat dissipation. In addition, dry ice can be used for cooling.
The package includes MultiDrive basic and a 250 ml milling chamber MI 250 with standard beater made of stainless steel. It is the perfect successor of the M 20 batch mill.