Hybridoma growth and expression CD media HYGM7
Supplier: BIOCONCEPT
Certificates
About this item
Specifications
- Sterility:Sterile
- Shipping conditions:Ambient
- Storage temperature:2…4 °C
Specifications
Product Information
- Catalog #Supplier #Shelf lifeSizePkAvailabilityPrice
- Hybridoma growth and expression CD media HYGM7 (liquid) without phenol redHybridoma growth and expression CD media HYGM7 (liquid) without phenol redBCON9-00F58-I9-00F58-I18 months500 ml bottle500 mL1 * 500 mL868,00 KčSupplier Article Number: 9-00F58-ILocal Article Number: BCON9-00F58-I
- Hybridoma growth and expression CD media HYGM7 (liquid) with phenol redHybridoma growth and expression CD media HYGM7 (liquid) with phenol redBCON9-00F67-I9-00F67-I18 months500 ml bottle500 mL1 * 500 mL868,00 KčSupplier Article Number: 9-00F67-ILocal Article Number: BCON9-00F67-I
- Hybridoma growth and expression CD media HYGM7 (liquid) with phenol redHybridoma growth and expression CD media HYGM7 (liquid) with phenol redBCON9-00F67-K9-00F67-K18 months1 l bottle1 L1 * 1 LSupplier Article Number: 9-00F67-KLocal Article Number: BCON9-00F67-K