Masterflex® Magnetic Drive Centrifugal Pump, Avantor®
- Environmentally Preferable:
- Model:
- 75226-10
- 75226-35
- 75226-32
- 75226-15
- 75226-12
- Duty cycle continuous:
- Yes
- Yes
- Yes
- Yes
- Yes
- Inlet connection:
- 1 NPT(F)
- 1,5 NPT(F)
- 1,5 NPT(F)
- 1 NPT(F)
- 1 NPT(F)
- Max. pressure:
- 7,6 bar (110 psi)
- 7,6 bar (110 psi)
- 7,6 bar (110 psi)
- 7,6 bar (110 psi)
- 7,6 bar (110 psi)
- Max. temperature:
- 71 °C (160 °F)
- 71 °C (160 °F)
- 71 °C (160 °F)
- 71 °C (160 °F)
- 71 °C (160 °F)
- Max. viscosity:
- 140 cP
- 140 cP
- 140 cP
- 140 cP
- 140 cP
- Motor type:
- TEFC
- TEFC
- TEFC
- TEFC
- Motor hp:
- 1
- 1
- 1/2
- 1/2
- Wetted materials:
- PP pump head and impeller, ceramic shaft, carbon fiber-filled PTFE thrust ring and sleeve bearings; EPDM (Nordel) O-ring
- PP pump head and impeller, ceramic shaft, carbon fiber-filled PTFE thrust ring and sleeve bearings; EPDM (Nordel) O-ring
- PP pump head and impeller, ceramic shaft, carbon fiber-filled PTFE thrust ring and sleeve bearings; EPDM (Nordel) O-ring
- PP pump head and impeller, ceramic shaft, carbon fiber-filled PTFE thrust ring and sleeve bearings; EPDM (Nordel) O-ring
- PP pump head and impeller, ceramic shaft, carbon fiber-filled PTFE thrust ring and sleeve bearings; EPDM (Nordel) O-ring
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- HeightLengthWidthPowerMaximum Flow Rate
- Mag Drive PP Centrifugal Pump Head; 32 gal./min/11,3 ftMag Drive PP Centrifugal Pump Head; 32 gal./min/11,3 ft75226-1017,5 cm17 cm16,8 cm121,1 L/min (32 gal./min)1 * 1 KS33 750,00 Kč
Specifications
Model75226-10Environmentally PreferableDuty cycle continuousYesInlet connection1 NPT(F)Max. pressure7,6 bar (110 psi)Max. temperature71 °C (160 °F)Max. viscosity140 cPMotor typeMotor hpWetted materialsPP pump head and impeller, ceramic shaft, carbon fiber-filled PTFE thrust ring and sleeve bearings; EPDM (Nordel) O-ringWarranty1 year - Mag Drive PP Centrifugal Pump; 32 gal./min/11,3 ft, 115/230VMag Drive PP Centrifugal Pump; 32 gal./min/11,3 ft, 115/230V75226-1222,9 cm39,4 cm22 cm115/230 VAC; 60 Hz; 6,4/3,2 A; 1 phase121,1 L/min (32 gal./min)1 * 1 KS46 240,00 KčCSA Certified
Specifications
Model75226-12Environmentally PreferableDuty cycle continuousYesInlet connection1 NPT(F)Max. pressure7,6 bar (110 psi)Max. temperature71 °C (160 °F)Max. viscosity140 cPMotor typeTEFCMotor hp1/2Wetted materialsPP pump head and impeller, ceramic shaft, carbon fiber-filled PTFE thrust ring and sleeve bearings; EPDM (Nordel) O-ringWarranty1 yearCSA Certified - Mag Drive PP Centrifugal Pump; 32 gal./min/11,3 ft, 230/460VMag Drive PP Centrifugal Pump; 32 gal./min/11,3 ft, 230/460V75226-1517,8 cm37,1 cm22 cm230/460 VAC; 60 Hz; 1,0/0,5 A; 3 phase121,1 L/min (32 gal./min)1 * 1 KS43 750,00 Kč
Specifications
Model75226-15Environmentally PreferableDuty cycle continuousYesInlet connection1 NPT(F)Max. pressure7,6 bar (110 psi)Max. temperature71 °C (160 °F)Max. viscosity140 cPMotor typeTEFCMotor hp1/2Wetted materialsPP pump head and impeller, ceramic shaft, carbon fiber-filled PTFE thrust ring and sleeve bearings; EPDM (Nordel) O-ringWarranty1 year - Mag Drive PP Centrifugal Pump; 68 gal./min/59 ft, 115/230VMag Drive PP Centrifugal Pump; 68 gal./min/59 ft, 115/230V75226-3222,9 cm44,7 cm22 cm115/230 VAC; 60 Hz; 12,0/6,0 A; 1 phase257,4 L/min (68 gal./min)1 * 1 KS55 740,00 Kč
Specifications
Model75226-32Environmentally PreferableDuty cycle continuousYesInlet connection1,5 NPT(F)Max. pressure7,6 bar (110 psi)Max. temperature71 °C (160 °F)Max. viscosity140 cPMotor typeTEFCMotor hp1Wetted materialsPP pump head and impeller, ceramic shaft, carbon fiber-filled PTFE thrust ring and sleeve bearings; EPDM (Nordel) O-ringWarranty1 year - Mag Drive PP Centrifugal Pump; 68 gal./min/59 ft, 230/460VMag Drive PP Centrifugal Pump; 68 gal./min/59 ft, 230/460V75226-3519,1 cm42,2 cm22 cm230/460 VAC; 60 Hz; 3,2/1,6 A; 3 phase257,4 L/min (68 gal./min)1 * 1 KS52 240,00 Kč
Specifications
Model75226-35Environmentally PreferableDuty cycle continuousYesInlet connection1,5 NPT(F)Max. pressure7,6 bar (110 psi)Max. temperature71 °C (160 °F)Max. viscosity140 cPMotor typeTEFCMotor hp1Wetted materialsPP pump head and impeller, ceramic shaft, carbon fiber-filled PTFE thrust ring and sleeve bearings; EPDM (Nordel) O-ringWarranty1 year