Order Entry
Masterflex® Liquid Flow Switches for High Inline Pressures, Avantor®
Masterflex® Liquid Flow Switches for High Inline Pressures, Avantor®
undefined
Masterflex® Liquid Flow Switches for High Inline Pressures, Avantor®
  • Model:
  • 32774-40
  • 32774-32
  • 32774-48
  • Max. pressure:
  • 1500 psi
  • 1500 psi
  • 1500 psi
  • Max. operating temperature:
  • 135 °C (275 °F)
  • 135 °C (275 °F)
  • 135 °C (275 °F)
  • Wetted materials:
  • Stainless Steel, PPS composite, 316 SS, and fluorocarbon
  • Brass, PPS composite, 316 SS, and fluorocarbon
  • Stainless Steel, PPS composite, 316 SS, and fluorocarbon
  • Process connection:
  • 0,375 NPT(M)
  • 0,375 NPT(M)
  • 0,375 NPT(M)
  • Height:
  • 40751"
  • 40751"
  • 40751"
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  •  
    Maximum Flow Rate
    Housing material
    Pk
  • Brass liquid flow switch for high inline pressures; 0,5 gal./min
    Brass liquid flow switch for high inline pressures; 0,5 gal./min
    32774-32
    0,5 gal./min
    Brass
    1
    1 * 1 KS
    1 * 1 KS - 8 130,00 Kč
    8 130,00 Kč
    1 * 1 KS
       8 130,00 Kč
     : 32774-32
     : MFLB32774-32
    Specifications
    Model
    32774-32
    Environmentally Preferable
    Max. pressure
    1500 psi
    Max. operating temperature
    135 °C (275 °F)
    Wetted materials
    Brass, PPS composite, 316 SS, and fluorocarbon
    Process connection
    0,375 NPT(M)
    Height
    40751"
    Warranty
    1 year
  • Stainless steel liquid flow switch for high inline pressures; 0,25 gal./min
    Stainless steel liquid flow switch for high inline pressures; 0,25 gal./min
    32774-40
    0,25 gal./min
    Stainless steel
    1
    1 * 1 KS
    1 * 1 KS - 18 280,00 Kč
    18 280,00 Kč
    1 * 1 KS
       18 280,00 Kč
     : 32774-40
     : MFLB32774-40
    Specifications
    Model
    32774-40
    Environmentally Preferable
    Max. pressure
    1500 psi
    Max. operating temperature
    135 °C (275 °F)
    Wetted materials
    Stainless Steel, PPS composite, 316 SS, and fluorocarbon
    Process connection
    0,375 NPT(M)
    Height
    40751"
    Warranty
    1 year
  • Stainless steel liquid flow switch for high inline pressures; 2,0 gal./min
    Stainless steel liquid flow switch for high inline pressures; 2,0 gal./min
    32774-48
    2 gal./min
    Stainless steel
    1
    1 * 1 KS
    1 * 1 KS - 18 950,00 Kč
    18 950,00 Kč
    1 * 1 KS
       18 950,00 Kč
     : 32774-48
     : MFLB32774-48
    Specifications
    Model
    32774-48
    Environmentally Preferable
    Max. pressure
    1500 psi
    Max. operating temperature
    135 °C (275 °F)
    Wetted materials
    Stainless Steel, PPS composite, 316 SS, and fluorocarbon
    Process connection
    0,375 NPT(M)
    Height
    40751"
    Warranty
    1 year