Masterflex® Sanitary Tri-Clamp® Screen Gaskets, EPDM, Avantor®
Supplier: Avantor Fluid Handling
Certificates
About this item
Protect your pump, valves and other components in fluid line.
- Pre-filter before critical membrane filters
- Generate turbulence for enhanced mass or heat transfer
- Purify in chromatography applications
Screen gaskets are designed to be interchangeable with standard sanitary clamp gaskets. Gaskets are available in platinum-cured silicone, Viton®, EPDM, PTFE, and Tuf-Steel in mesh sizes of 10, 20, 40, 60 and 100. Screen mesh is constructed of 316L SS. Gaskets meet USP Class VI, USDA, 3-A and cGMP criteria. FDA compliant. All gaskets are animal-derived ingredient free. Each gasket sold individually.
EPDM gaskets offer an economical solution to transfer water, acetones, and ketones. Gaskets are ideal in chemical process applications; temperature range is –65 to 212 °F (–54 to 100 °C).
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX30537-82
- MFLX30537-20
- MFLX30537-84
- MFLX30537-08
- MFLX30537-66
- MFLX30537-24
- MFLX30537-80
- MFLX30537-22
- MFLX30537-62
- MFLX30537-48
- MFLX30537-64
- MFLX30537-44
- MFLX30537-04
- MFLX30537-68
- MFLX30537-46
- MFLX30537-60
- MFLX30537-42
- MFLX30537-40
- MFLX30537-06
- MFLX30537-26
- MFLX30537-86
- MFLX30537-88
- MFLX30537-28
- Connection type:
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
Specifications
Product Information
- Catalog #Supplier #Fitting materialMesh sizeParticle size retainedSanitary clamp size (Port 1)Sanitary gasket sizePkAvailabilityPrice
- EPDM Screen Sanitary Gasket, Size 100 SS Mesh, 3/4"; 1/EAEPDM Screen Sanitary Gasket, Size 100 SS Mesh, 3/4"; 1/EA-EPDM100140 µm3/4"3/4"1Log in for availability1 * 1 KS1 660,00 KčSpecifications:
More
Specifications
Connection typeSanitaryEnvironmentally PreferableWarranty1 year - EPDM Screen Sanitary Gasket, Size 20 SS Mesh, 1/2" ; 1/PkEPDM Screen Sanitary Gasket, Size 20 SS Mesh, 1/2" ; 1/Pk30537-20EPDM20864 µm3/4"0,5"1Log in for availability1 * 1 KS1 900,00 KčSpecifications:
More
Specifications
Connection typeSanitaryEnvironmentally PreferableWarranty1 year - EPDM Screen Sanitary Gasket, Size 100 SS Mesh, 1"; 1/EAEPDM Screen Sanitary Gasket, Size 100 SS Mesh, 1"; 1/EA-EPDM100140 µm1-1/2"1"1Log in for availability1 * 1 KS1 890,00 KčSpecifications:
More
Specifications
Connection typeSanitaryEnvironmentally PreferableWarranty1 year - EPDM Screen Sanitary Gasket, Size 10 SS Mesh, 2" ; 1/PkEPDM Screen Sanitary Gasket, Size 10 SS Mesh, 2" ; 1/Pk30537-08EPDM101905 µm2"2"1Log in for availability1 * 1 KS1 900,00 KčSpecifications:
More
Specifications
Connection typeSanitaryEnvironmentally PreferableWarranty1 year - EPDM Screen Sanitary Gasket, Size 60 SS Mesh, 1-1/2"; 1/EAEPDM Screen Sanitary Gasket, Size 60 SS Mesh, 1-1/2"; 1/EA-EPDM100233 µm1-1/2"1-1/2"1Log in for availability1 * 1 KS1 890,00 KčSpecifications:
More
Specifications
Connection typeSanitaryEnvironmentally PreferableWarranty1 year - EPDM Screen Sanitary Gasket, Size 20 SS Mesh, 1" ; 1/PkEPDM Screen Sanitary Gasket, Size 20 SS Mesh, 1" ; 1/Pk30537-24EPDM20864 µm1-1/2"1"1Log in for availability1 * 1 KS1 890,00 KčSpecifications:
More
Specifications
Connection typeSanitaryEnvironmentally PreferableWarranty1 year - EPDM Screen Sanitary Gasket, Size 100 SS Mesh, 1/2"; 1/EAEPDM Screen Sanitary Gasket, Size 100 SS Mesh, 1/2"; 1/EA-EPDM100140 µm3/4"1/2"1Log in for availability1 * 1 KS1 660,00 KčSpecifications:
More
Specifications
Connection typeSanitaryEnvironmentally PreferableWarranty1 year - EPDM Screen Sanitary Gasket, Size 20 SS Mesh, 3/4" ; 1/PkEPDM Screen Sanitary Gasket, Size 20 SS Mesh, 3/4" ; 1/Pk30537-22EPDM20864 µm3/4"0,75"1Log in for availability1 * 1 KS1 900,00 KčSpecifications:
More
Specifications
Connection typeSanitaryEnvironmentally PreferableWarranty1 year - EPDM Screen Sanitary Gasket, Size 60 SS Mesh, 3/4"; 1/EAEPDM Screen Sanitary Gasket, Size 60 SS Mesh, 3/4"; 1/EA-EPDM100233 µm3/4"3/4"1Log in for availability1 * 1 KS1 710,00 KčSpecifications:
More
Specifications
Connection typeSanitaryEnvironmentally PreferableWarranty1 year - EPDM Screen Sanitary Gasket, Size 40 SS Mesh, 2" ; 1/PkEPDM Screen Sanitary Gasket, Size 40 SS Mesh, 2" ; 1/Pk30537-48EPDM40381 µm2"2"1Log in for availability1 * 1 KS1 780,00 KčSpecifications:
More
Specifications
Connection typeSanitaryEnvironmentally PreferableWarranty1 year - EPDM Screen Sanitary Gasket, Size 60 SS Mesh, 1"; 1/EAEPDM Screen Sanitary Gasket, Size 60 SS Mesh, 1"; 1/EA-EPDM100233 µm1-1/2"1"1Log in for availability1 * 1 KS1 890,00 KčSpecifications:
More
Specifications
Connection typeSanitaryEnvironmentally PreferableWarranty1 year - EPDM Screen Sanitary Gasket, Size 40 SS Mesh, 1" ; 1/PkEPDM Screen Sanitary Gasket, Size 40 SS Mesh, 1" ; 1/Pk30537-44EPDM40381 µm1-1/2"1"1Log in for availability1 * 1 KS1 750,00 KčSpecifications:
More
Specifications
Connection typeSanitaryEnvironmentally PreferableWarranty1 year - EPDM Screen Sanitary Gasket, Size 10 SS Mesh, 1" ; 1/PkEPDM Screen Sanitary Gasket, Size 10 SS Mesh, 1" ; 1/Pk30537-04EPDM101905 µm1-1/2"1"1Log in for availability1 * 1 KS1 720,00 KčSpecifications:
More
Specifications
Connection typeSanitaryEnvironmentally PreferableWarranty1 year - EPDM Screen Sanitary Gasket, Size 60 SS Mesh, 2" ; 1/PkEPDM Screen Sanitary Gasket, Size 60 SS Mesh, 2" ; 1/Pk30537-68EPDM60233 µm2"2"1Log in for availability1 * 1 KS1 970,00 KčSpecifications:
More
Specifications
Connection typeSanitaryEnvironmentally PreferableWarranty1 year - EPDM Screen Sanitary Gasket, Size 40 SS Mesh, 1-1/2"; 1/EAEPDM Screen Sanitary Gasket, Size 40 SS Mesh, 1-1/2"; 1/EA-EPDM60381 µm1-1/2"1-1/2"1Log in for availability1 * 1 KS1 890,00 KčSpecifications:
More
Specifications
Connection typeSanitaryEnvironmentally PreferableWarranty1 year - EPDM Screen Sanitary Gasket, Size 60 SS Mesh, 1/2" ; 1/PkEPDM Screen Sanitary Gasket, Size 60 SS Mesh, 1/2" ; 1/Pk30537-60EPDM60233 µm3/4"0,5"1Log in for availability1 * 1 KS1 710,00 KčSpecifications:
More
Specifications
Connection typeSanitaryEnvironmentally PreferableWarranty1 year - EPDM Screen Sanitary Gasket, Size 40 SS Mesh, 3/4" ; 1/PkEPDM Screen Sanitary Gasket, Size 40 SS Mesh, 3/4" ; 1/Pk30537-42EPDM40381 µm3/4"0,75"1Log in for availability1 * 1 KS1 710,00 KčSpecifications:
More
Specifications
Connection typeSanitaryEnvironmentally PreferableWarranty1 year - EPDM Screen Sanitary Gasket, Size 40 SS Mesh, 1/2" ; 1/PkEPDM Screen Sanitary Gasket, Size 40 SS Mesh, 1/2" ; 1/Pk30537-40EPDM40381 µm3/4"0,5"1Log in for availability1 * 1 KS1 710,00 KčSpecifications:
More
Specifications
Connection typeSanitaryEnvironmentally PreferableWarranty1 year - EPDM Screen Sanitary Gasket, Size 10 SS Mesh, 1-1/2"; 1/EAEPDM Screen Sanitary Gasket, Size 10 SS Mesh, 1-1/2"; 1/EA-EPDM601905 µm1-1/2"1-1/2"1Log in for availability1 * 1 KS1 720,00 KčSpecifications:
More
Specifications
Connection typeSanitaryEnvironmentally PreferableWarranty1 year - EPDM Screen Sanitary Gasket, Size 20 SS Mesh, 1-1/2"; 1/EAEPDM Screen Sanitary Gasket, Size 20 SS Mesh, 1-1/2"; 1/EA-EPDM60864 µm1-1/2"1-1/2"1Log in for availability1 * 1 KS1 890,00 KčSpecifications:
More
Specifications
Connection typeSanitaryEnvironmentally PreferableWarranty1 year - EPDM Screen Sanitary Gasket, Size 100 SS Mesh, 1-1/2"; 1/EAEPDM Screen Sanitary Gasket, Size 100 SS Mesh, 1-1/2"; 1/EA-EPDM100140 µm1-1/2"1-1/2"1Log in for availability1 * 1 KS1 890,00 KčSpecifications:
More
Specifications
Connection typeSanitaryEnvironmentally PreferableWarranty1 year - EPDM Screen Sanitary Gasket, Size 100 SS Mesh, 2"; 1/EAEPDM Screen Sanitary Gasket, Size 100 SS Mesh, 2"; 1/EA-EPDM20140 µm2"2"1Log in for availability1 * 1 KS1 970,00 KčSpecifications:
More
Specifications
Connection typeSanitaryEnvironmentally PreferableWarranty1 year - EPDM Screen Sanitary Gasket, Size 20 SS Mesh, 2" ; 1/PkEPDM Screen Sanitary Gasket, Size 20 SS Mesh, 2" ; 1/Pk30537-28EPDM20864 µm2"2"1Log in for availability1 * 1 KS1 720,00 KčSpecifications:
More
Specifications
Connection typeSanitaryEnvironmentally PreferableWarranty1 year