GF Signet® Paddle-Wheel Flow Sensors
Easily configure to your needs.
- Built-in user interface for on-site configuration
- Low power and high resolution
Fluid measurement with paddlewheel sensors
Digital (S3L) or 4 to 20mA outputs, or Flow Switch, or Pulse output (multi-function)
Allows for up to six sensors to Signet 8900 Controller
Low flow capabilities down to 0.1 m/s (0.3 ft/s)
Polypropylene or PVDF wetted materials
- Environmentally Preferable:
- Input:
- 1 to 1000 Hz Frequency
- 1 to 1000 Hz Frequency
- 1 to 1000 Hz Frequency
- 1 to 1000 Hz Frequency
- Min. operating temperature:
- –18 °C (0 °F)
- –18 °C (0 °F)
- –18 °C (0 °F)
- –18 °C (0 °F)
- Max. operating temperature:
- 85 °C (185 °F)
- 85 °C (185 °F)
- 85 °C (185 °F)
- 85 °C (185 °F)
- Max. pressure:
- 180 psi
- 180 psi
- 200 psi
- 180 psi
- O-Ring material:
- FPM
- FPM
- FPM
- FPM
- Wetted materials:
- Polypropylene body, black PVDF rotor, Titanium pin, FPM O-rings
- Polypropylene body, black PVDF rotor, Titanium pin, FPM O-rings
- Natural PVDF body, rotor and pin, FPM O-rings
- Polypropylene body, black PVDF rotor, Titanium pin, FPM O-rings
- Warranty:
- 2 years
- 2 years
- 2 years
- 2 years
- Fluid typeHousing materialOutputPipe sizePk
- Paddlewheel Flow Sensor, Pulse Divider/Dry Contact, Polypropylene, 0,5" to 4" Pipe SizePaddlewheel Flow Sensor, Pulse Divider/Dry Contact, Polypropylene, 0,5" to 4" Pipe Size32512-00LiquidsPPPulse divider/dry contact relay½" to 4" diameter11 * 1 KS26 750,00 Kč
Specifications
Environmentally PreferableInput1 to 1000 Hz FrequencyMin. operating temperature–18 °C (0 °F)Max. operating temperature85 °C (185 °F)Max. pressure180 psiO-Ring materialFPMWetted materialsPolypropylene body, black PVDF rotor, Titanium pin, FPM O-ringsWarranty2 years - Paddlewheel Flow Sensor, 4-20 mA Output, Polypropylene, 0,5" to 4" Pipe SizePaddlewheel Flow Sensor, 4-20 mA Output, Polypropylene, 0,5" to 4" Pipe Size32512-15LiquidsPP4 to 20 mA½" to 4" diameter11 * 1 KS29 250,00 Kč
Specifications
Environmentally PreferableInput1 to 1000 Hz FrequencyMin. operating temperature–18 °C (0 °F)Max. operating temperature85 °C (185 °F)Max. pressure180 psiO-Ring materialFPMWetted materialsPolypropylene body, black PVDF rotor, Titanium pin, FPM O-ringsWarranty2 years - Paddlewheel Flow Sensor, 4-20 mA Output, PVDF, 0,5" to 4" Pipe SizePaddlewheel Flow Sensor, 4-20 mA Output, PVDF, 0,5" to 4" Pipe Size32512-17LiquidsPVDF4 to 20 mA½" to 4" diameter11 * 1 KS35 000,00 Kč
Specifications
Environmentally PreferableInput1 to 1000 Hz FrequencyMin. operating temperature–18 °C (0 °F)Max. operating temperature85 °C (185 °F)Max. pressure200 psiO-Ring materialFPMWetted materialsNatural PVDF body, rotor and pin, FPM O-ringsWarranty2 years - Paddlewheel Flow Sensor, 4-20 mA Output, Polypropylene, 5" to 8" Pipe SizePaddlewheel Flow Sensor, 4-20 mA Output, Polypropylene, 5" to 8" Pipe Size32512-16LiquidsPP4 to 20 mA5" to 8" diameter11 * 1 KS30 500,00 Kč
Specifications
Environmentally PreferableInput1 to 1000 Hz FrequencyMin. operating temperature–18 °C (0 °F)Max. operating temperature85 °C (185 °F)Max. pressure180 psiO-Ring materialFPMWetted materialsPolypropylene body, black PVDF rotor, Titanium pin, FPM O-ringsWarranty2 years