Order Entry
Masterflex® Adapter Fittings, Compression to Male Threaded, Elbows, Avantor®
Masterflex® Adapter Fittings, Compression to Male Threaded, Elbows, Avantor®
undefined
Masterflex® Adapter Fittings, Compression to Male Threaded, Elbows, Avantor®
  • Model:
  • 06386-10
  • 06386-50
  • Orifice dimensions:
  • 6,35 mm (0,25")
  • 8,73 mm (0,34375")
  • Sterilisation:
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  •  
    Connection Type
    Fitting material
    Flow pattern
    Thread size NPT(M) (port 2)
    Pk
  • Compression male pipe adapter elbow, ¼" × ⅛", polypropylene, 10 Pack
    Compression male pipe adapter elbow, ¼" × ⅛", polypropylene, 10 Pack
    06386-10
    Compression to Thread
    Polypropylene
    Elbow
    ⅛"
    10
    1 * 10 KS
    1 * 10 KS - 675,00 Kč
    675,00 Kč
    1 * 10 KS
       675,00 Kč
     : 06386-10
     : MFLB06386-10
    Specifications
    Model
    06386-10
    Environmentally Preferable
    Orifice dimensions
    6,35 mm (0,25")
    Sterilisation
    Ethylene oxide (ETO) only
  • Compression male pipe adapter elbow, ⅜" × ⅜", polypropylene, 10 Pack
    Compression male pipe adapter elbow, ⅜" × ⅜", polypropylene, 10 Pack
    06386-50
    Compression to Thread
    Polypropylene
    Elbow
    ⅜"
    10
    1 * 10 KS
    1 * 10 KS - 1 770,00 Kč
    1 770,00 Kč
    1 * 10 KS
       1 770,00 Kč
     : 06386-50
     : MFLB06386-50
    Specifications
    Model
    06386-50
    Environmentally Preferable
    Orifice dimensions
    8,73 mm (0,34375")
    Sterilisation
    Ethylene oxide (ETO) only