Masterflex® Transfer Tubing, Tygon® 2475 I.B., High Purity Reinforced, Avantor®
Supplier: Avantor Fluid Handling
Certificates
About this item
Keeps visual track of samples.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimise downtime
- Regulatory approvals to match your unique application
Formulation: TYGON® 2475 I.B.
Applications: Pharmaceutical and cosmetic processing, deionized water systems, cell harvest and media process systems, chemical transfer, preservative and sterile fill lines.
Characteristics: Plasticizer free, smooth inner surface (inhibits particulate build-up and bacterial growth), and nontoxic. Transparent.
Certification: Resin is FDA-compliant (21 CFR 177.1550) and meets NSF criteria. Tubing is manufactured according to GMP.
Temperature range: -94 to 125 °F (-70 to 52 °C)
Sterilize: With ethylene oxide or gamma irradiation.
- Environmentally Preferable:
Specifications
- Model:
- 95360-50
- 95360-51
- 95360-52
- 95360-53
- 95360-55
- 95360-54
- Bend radius:
- 9,5 mm (0,375")
- 25,4 mm (1")
- 25,4 mm (1")
- 44,5 mm (1,75")
- 88,9 mm (3,5")
- 31,8 mm (1,25")
- Clarity:
- Clear
- Clear
- Clear
- Clear
- Clear
- Clear
- Length per pack:
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 7,62 m (25 ft.)
- 15,2 m (50 ft.)
- Max. pressure:
- 15,5 bar (225 psi)
- 14,5 bar (210 psi)
- 15,9 bar (230 psi)
- 9,3 bar (135 psi)
- 8,6 bar (125 psi)
- 9,3 bar (135 psi)
- Max. temperature:
- 52 °C (125 °F)
- 52 °C (125 °F)
- 52 °C (125 °F)
- 52 °C (125 °F)
- 52 °C (125 °F)
- 52 °C (125 °F)
- Max. vacuum:
- 760 mm Hg (29,9" Hg)
- 760 mm Hg (29,9" Hg)
- 760 mm Hg (29,9" Hg)
- 760 mm Hg (29,9" Hg)
- 760 mm Hg (29,9" Hg)
- 760 mm Hg (29,9" Hg)
- Min. temperature:
- –70 °C (–94 °F)
- –70 °C (–94 °F)
- –70 °C (–94 °F)
- –70 °C (–94 °F)
- –70 °C (–94 °F)
- –70 °C (–94 °F)
- Regulatory approvals:
- USP
- USP
- USP
- USP
- USP
- USP
- Sterilisation method autoclave:
- No
- No
- No
- No
- No
- No
- Sterilisation method EtO:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Sterilisation method gamma:
- Yes, up to 45 kGy
- Yes, up to 45 kGy
- Yes, up to 45 kGy
- Yes, up to 45 kGy
- Yes, up to 45 kGy
- Yes, up to 45 kGy
- Working pressure:
- 15,5 bar (225 psi)
- 14,5 bar (210 psi)
- 15,9 bar (230 psi)
- 9,3 bar (135 psi)
- 8,6 bar (125 psi)
- 9,3 bar (135 psi)
Specifications
Product Information
- Catalog #Supplier #Tubing int.ØWall thicknessTubing ext.ØTubing materialLengthAvailabilityPrice
- Transfer Tubing, Tygon® 2475 I.B. Reinforced, 1/2" ID x 3/4" OD; 50 FtTransfer Tubing, Tygon® 2475 I.B. Reinforced, 1/2" ID x 3/4" OD; 50 Ft95360-5212,8 mm (0,5")3,2 mm19,1 mm (0,75")Tygon® 2475 I.B.15,2 mLog in for availability1 * 50 Feet22 180,00 KčSpecifications:
More
Specifications
Model95360-52Environmentally PreferableBend radius25,4 mm (1")ClarityClearLength per pack15,2 m (50 ft.)Max. pressure15,9 bar (230 psi)Max. temperature52 °C (125 °F)Max. vacuum760 mm Hg (29,9" Hg)Min. temperature–70 °C (–94 °F)Regulatory approvalsUSPSterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaYes, up to 45 kGyWorking pressure15,9 bar (230 psi) - Transfer Tubing, Tygon® 2475 I.B. Reinforced, 1/4" ID x 1/2" OD; 50 FtTransfer Tubing, Tygon® 2475 I.B. Reinforced, 1/4" ID x 1/2" OD; 50 Ft95360-506,4 mm (0,25")3,2 mm12,7 mm (0,5")Tygon® 2475 I.B.15,2 mLog in for availability1 * 50 Feet13 150,00 KčSpecifications:
More
Specifications
Model95360-50Environmentally PreferableBend radius9,5 mm (0,375")ClarityClearLength per pack15,2 m (50 ft.)Max. pressure15,5 bar (225 psi)Max. temperature52 °C (125 °F)Max. vacuum760 mm Hg (29,9" Hg)Min. temperature–70 °C (–94 °F)Regulatory approvalsUSPSterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaYes, up to 45 kGyWorking pressure15,5 bar (225 psi) - Transfer Tubing, Tygon® 2475 I.B. Reinforced, 1" ID x 1-3/8" OD; 25 FtTransfer Tubing, Tygon® 2475 I.B. Reinforced, 1" ID x 1-3/8" OD; 25 Ft95360-5525,4 mm (1")4,8 mm35 mm (1,375")Tygon® 2475 I.B.7,62 mLog in for availability1 * 25 Feet24 320,00 KčSpecifications:
More
Specifications
Model95360-55Environmentally PreferableBend radius88,9 mm (3,5")ClarityClearLength per pack7,62 m (25 ft.)Max. pressure8,6 bar (125 psi)Max. temperature52 °C (125 °F)Max. vacuum760 mm Hg (29,9" Hg)Min. temperature–70 °C (–94 °F)Regulatory approvalsUSPSterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaYes, up to 45 kGyWorking pressure8,6 bar (125 psi) - Transfer Tubing, Tygon® 2475 I.B. Reinforced, 3/4" ID x 1-1/16" OD; 50 FtTransfer Tubing, Tygon® 2475 I.B. Reinforced, 3/4" ID x 1-1/16" OD; 50 Ft95360-5419,1 mm (0,75")9,6 mm27 mm (1,0625")Tygon® 2475 I.B.15,2 mLog in for availability1 * 50 Feet28 500,00 KčSpecifications:
More
Specifications
Model95360-54Environmentally PreferableBend radius31,8 mm (1,25")ClarityClearLength per pack15,2 m (50 ft.)Max. pressure9,3 bar (135 psi)Max. temperature52 °C (125 °F)Max. vacuum760 mm Hg (29,9" Hg)Min. temperature–70 °C (–94 °F)Regulatory approvalsUSPSterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaYes, up to 45 kGyWorking pressure9,3 bar (135 psi) - Transfer Tubing, Tygon® 2475 I.B. Reinforced, 3/8" ID x 5/8" OD; 50 FtTransfer Tubing, Tygon® 2475 I.B. Reinforced, 3/8" ID x 5/8" OD; 50 Ft95360-519,5 mm (0,375")3,2 mm16 mm (0,625")Tygon® 2475 I.B.15,2 mLog in for availability1 * 50 Feet15 600,00 KčSpecifications:
More
Specifications
Model95360-51Environmentally PreferableBend radius25,4 mm (1")ClarityClearLength per pack15,2 m (50 ft.)Max. pressure14,5 bar (210 psi)Max. temperature52 °C (125 °F)Max. vacuum760 mm Hg (29,9" Hg)Min. temperature–70 °C (–94 °F)Regulatory approvalsUSPSterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaYes, up to 45 kGyWorking pressure14,5 bar (210 psi) - Transfer Tubing, Tygon® 2475 I.B. Reinforced, 5/8" ID x 7/8" OD; 50 FtTransfer Tubing, Tygon® 2475 I.B. Reinforced, 5/8" ID x 7/8" OD; 50 Ft95360-5316 mm (0,625")3,2 mm22,2 mm (0,875")Tygon® 2475 I.B.15,2 mLog in for availability1 * 50 Feet26 000,00 KčSpecifications:
More
Specifications
Model95360-53Environmentally PreferableBend radius44,5 mm (1,75")ClarityClearLength per pack15,2 m (50 ft.)Max. pressure9,3 bar (135 psi)Max. temperature52 °C (125 °F)Max. vacuum760 mm Hg (29,9" Hg)Min. temperature–70 °C (–94 °F)Regulatory approvalsUSPSterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaYes, up to 45 kGyWorking pressure9,3 bar (135 psi)