Order Entry
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Puri-Clear™ LL, Avantor®
Certificates
undefined
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Puri-Clear™ LL, Avantor®
Certificates
Specifications
  • Model:
  • 95625-48
  • 95625-16
  • 95625-26
  • 95625-24
  • 95625-22
  • 95625-18
  • 95625-10
  • 95625-34
  • 95625-32
  • 95625-30
  • 95625-28
  • 95625-42
  • 95625-40
  • 95625-14
  • 95625-38
  • 95625-12
  • 95625-36
  • 95625-46
  • 95625-44
  • 95699-39
  • Regulatory approvals:
  • ADCF;Non-DEHP;RoHS;USP
  • ADCF;Non-DEHP;RoHS;USP
  • ADCF;Non-DEHP;RoHS;USP
  • ADCF;Non-DEHP;RoHS;USP
  • ADCF;Non-DEHP;RoHS;USP
  • ADCF;Non-DEHP;RoHS;USP
  • ADCF;Non-DEHP;RoHS;USP
  • ADCF;Non-DEHP;RoHS;USP
  • ADCF;Non-DEHP;RoHS;USP
  • ADCF;Non-DEHP;RoHS;USP
  • ADCF;Non-DEHP;RoHS;USP
  • ADCF;Non-DEHP;RoHS;USP
  • ADCF;Non-DEHP;RoHS;USP
  • ADCF;Non-DEHP;RoHS;USP
  • ADCF;Non-DEHP;RoHS;USP
  • ADCF;Non-DEHP;RoHS;USP
  • ADCF;Non-DEHP;RoHS;USP
  • ADCF;Non-DEHP;RoHS;USP
  • ADCF;Non-DEHP;RoHS;USP
  • ADCF;Non-DEHP;RoHS;USP
  • Series:
  • Similar to SC0052-AI
  • Similar to SC0057-AI
  • Similar to SC0056-AI
  • Similar to SC0055-AI
  • Similar to SC0053-AI
  • Similar to SC0049-AI
  • Similar to SC0064-AI
  • Similar to SC0063-AI
  • Similar to SC0061-AI
  • Similar to SC0059-AI
  • Similar to SC0069-AI
  • Similar to SC0068-AI
  • Similar to SC0051-AI
  • Similar to SC0066-AI
  • Similar to SC0050-AI
  • Similar to SC0065-AI
  • Similar to SC0071-AI
  • Similar to SC0070-AI
  • Tubes per pack:
  • 12
  • 12
  • 12
  • 12
  • 12
  • 12
  • 12
  • 12
  • 12
  • 12
  • 12
  • 12
  • 12
  • 12
  • 12
  • 12
  • 12
  • 12
  • 12
  • 12
Specifications
Related Information
Product Information
  • Catalog #
    Tubing int.Ø
    Colour coded stops
    Length
    Tubing material
    Pk
    Availability
    Price
  • Pump Tubing, 3-Stop, Puri-Clear™ LL, 0,19 mm ID; 12/PK
    Pump Tubing, 3-Stop, Puri-Clear™ LL, 0,19 mm ID; 12/PK
    MFLX95625-10
    0,19 mm
    Orange/red/orange
    406 mm
    Puri-Clear™ LL
    12
    Log in for availability
    1 * 12 KS
    1 * 12 KS - 1 460,00 Kč
    1 460,00 Kč
    1 * 12 KS
       1 460,00 Kč
     : MFLA95625-10
    Specifications:
    More
    Specifications
    Model
    95625-10
    Environmentally Preferable
    Regulatory approvals
    ADCF;Non-DEHP;RoHS;USP
    Series
    Similar to SC0049-AI
    Tubes per pack
    12
  • Pump Tubing, 3-Stop, Puri-Clear™ LL, 0,25 mm ID; 12/PK
    Pump Tubing, 3-Stop, Puri-Clear™ LL, 0,25 mm ID; 12/PK
    0,25 mm
    Orange/blue/orange
    455 mm
    Puri-Clear™ LL
    12
    1 * 12 KS
    1 * 12 KS - 1 460,00 Kč
    1 460,00 Kč
    1 * 12 KS
       1 460,00 Kč
     : MFLA95625-12
    Specifications
    Model
    95625-12
    Environmentally Preferable
    Regulatory approvals
    ADCF;Non-DEHP;RoHS;USP
    Series
    Similar to SC0050-AI
    Tubes per pack
    12
  • Pump Tubing, 3-Stop, Puri-Clear™ LL, 0,38 mm ID; 12/PK
    Pump Tubing, 3-Stop, Puri-Clear™ LL, 0,38 mm ID; 12/PK
    0,38 mm
    Orange/green/orange
    406 mm
    Puri-Clear™ LL
    12
    1 * 12 KS
    1 * 12 KS - 1 460,00 Kč
    1 460,00 Kč
    1 * 12 KS
       1 460,00 Kč
     : MFLA95625-14
    Specifications
    Model
    95625-14
    Environmentally Preferable
    Regulatory approvals
    ADCF;Non-DEHP;RoHS;USP
    Series
    Similar to SC0051-AI
    Tubes per pack
    12
  • Pump Tubing, 3-Stop, Puri-Clear™ LL, 0,44 mm ID; 12/PK
    Pump Tubing, 3-Stop, Puri-Clear™ LL, 0,44 mm ID; 12/PK
    0,44 mm
    Green/Yellow/green
    406 mm
    Puri-Clear™ LL
    12
    1 * 12 KS
    1 * 12 KS - 1 460,00 Kč
    1 460,00 Kč
    1 * 12 KS
       1 460,00 Kč
     : MFLA95625-16
    Specifications
    Model
    95625-16
    Environmentally Preferable
    Regulatory approvals
    ADCF;Non-DEHP;RoHS;USP
    Series
    Similar to SC0052-AI
    Tubes per pack
    12
  • Pump Tubing, 3-Stop, Puri-Clear™ LL, 0,51 mm ID; 12/PK
    Pump Tubing, 3-Stop, Puri-Clear™ LL, 0,51 mm ID; 12/PK
    0,51 mm
    Orange/yellow/orange
    406 mm
    Puri-Clear™ LL
    12
    1 * 12 KS
    1 * 12 KS - 1 460,00 Kč
    1 460,00 Kč
    1 * 12 KS
       1 460,00 Kč
     : MFLA95625-18
    Specifications
    Model
    95625-18
    Environmentally Preferable
    Regulatory approvals
    ADCF;Non-DEHP;RoHS;USP
    Series
    Similar to SC0053-AI
    Tubes per pack
    12
  • Pump Tubing, 3-Stop, Puri-Clear™ LL, 0,64 mm ID; 12/PK
    Pump Tubing, 3-Stop, Puri-Clear™ LL, 0,64 mm ID; 12/PK
    0,64 mm
    Orange/white/orange
    406 mm
    Puri-Clear™ LL
    12
    1 * 12 KS
    1 * 12 KS - 1 460,00 Kč
    1 460,00 Kč
    1 * 12 KS
       1 460,00 Kč
     : MFLA95625-22
    Specifications
    Model
    95625-22
    Environmentally Preferable
    Regulatory approvals
    ADCF;Non-DEHP;RoHS;USP
    Series
    Similar to SC0055-AI
    Tubes per pack
    12
  • Pump Tubing, 3-Stop, Puri-Clear™ LL, 0,76 mm ID; 12/PK
    Pump Tubing, 3-Stop, Puri-Clear™ LL, 0,76 mm ID; 12/PK
    0,76 mm
    Black/Black/black
    406 mm
    Puri-Clear™ LL
    12
    1 * 12 KS
    1 * 12 KS - 1 460,00 Kč
    1 460,00 Kč
    1 * 12 KS
       1 460,00 Kč
     : MFLA95625-24
    Specifications
    Model
    95625-24
    Environmentally Preferable
    Regulatory approvals
    ADCF;Non-DEHP;RoHS;USP
    Series
    Similar to SC0056-AI
    Tubes per pack
    12
  • Pump Tubing, 3-Stop, Puri-Clear™ LL, 0,89 mm ID; 12/PK
    Pump Tubing, 3-Stop, Puri-Clear™ LL, 0,89 mm ID; 12/PK
    0,89 mm
    Orange/orange/orange
    406 mm
    Puri-Clear™ LL
    12
    1 * 12 KS
    1 * 12 KS - 1 460,00 Kč
    1 460,00 Kč
    1 * 12 KS
       1 460,00 Kč
     : MFLA95625-26
    Specifications
    Model
    95625-26
    Environmentally Preferable
    Regulatory approvals
    ADCF;Non-DEHP;RoHS;USP
    Series
    Similar to SC0057-AI
    Tubes per pack
    12
  • Pump Tubing, 3-Stop, Puri-Clear™ LL, 1,02 mm ID; 12/PK
    Pump Tubing, 3-Stop, Puri-Clear™ LL, 1,02 mm ID; 12/PK
    1,02 mm
    White/white/white
    406 mm
    Puri-Clear™ LL
    12
    1 * 12 KS
    1 * 12 KS - 1 460,00 Kč
    1 460,00 Kč
    1 * 12 KS
       1 460,00 Kč
     : MFLA95625-28
    Specifications
    Model
    95625-28
    Environmentally Preferable
    Regulatory approvals
    ADCF;Non-DEHP;RoHS;USP
    Series
    Similar to SC0059-AI
    Tubes per pack
    12
  • Pump Tubing, 3-Stop, Puri-Clear™ LL, 1,14 mm ID; 12/PK
    Pump Tubing, 3-Stop, Puri-Clear™ LL, 1,14 mm ID; 12/PK
    1,14 mm
    Red/Red/red
    406 mm
    Puri-Clear™ LL
    12
    1 * 12 KS
    1 * 12 KS - 1 460,00 Kč
    1 460,00 Kč
    1 * 12 KS
       1 460,00 Kč
     : MFLA95625-30
    Specifications
    Model
    95625-30
    Environmentally Preferable
    Regulatory approvals
    ADCF;Non-DEHP;RoHS;USP
    Series
    Similar to SC0061-AI
    Tubes per pack
    12
  • Pump Tubing, 3-Stop, Puri-Clear™ LL, 1,30 mm ID; 12/PK
    Pump Tubing, 3-Stop, Puri-Clear™ LL, 1,30 mm ID; 12/PK
    1,3 mm
    Gray/Gray/Gray
    406 mm
    Puri-Clear™ LL
    12
    1 * 12 KS
    1 * 12 KS - 1 460,00 Kč
    1 460,00 Kč
    1 * 12 KS
       1 460,00 Kč
     : MFLA95625-32
    Specifications
    Model
    95625-32
    Environmentally Preferable
    Regulatory approvals
    ADCF;Non-DEHP;RoHS;USP
    Series
    Similar to SC0063-AI
    Tubes per pack
    12
  • Pump Tubing, 3-Stop, Puri-Clear™ LL, 1,42 mm ID; 12/PK
    Pump Tubing, 3-Stop, Puri-Clear™ LL, 1,42 mm ID; 12/PK
    1,42 mm
    Yellow/yellow/yellow
    406 mm
    Puri-Clear™ LL
    12
    1 * 12 KS
    1 * 12 KS - 1 460,00 Kč
    1 460,00 Kč
    1 * 12 KS
       1 460,00 Kč
     : MFLA95625-34
    Specifications
    Model
    95625-34
    Environmentally Preferable
    Regulatory approvals
    ADCF;Non-DEHP;RoHS;USP
    Series
    Similar to SC0064-AI
    Tubes per pack
    12
  • Pump Tubing, 3-Stop, Puri-Clear™ LL, 1,52 mm ID; 12/PK
    Pump Tubing, 3-Stop, Puri-Clear™ LL, 1,52 mm ID; 12/PK
    1,52 mm
    Yellow/blue/yellow
    406 mm
    Puri-Clear™ LL
    12
    1 * 12 KS
    1 * 12 KS - 1 460,00 Kč
    1 460,00 Kč
    1 * 12 KS
       1 460,00 Kč
     : MFLA95625-36
    Specifications
    Model
    95625-36
    Environmentally Preferable
    Regulatory approvals
    ADCF;Non-DEHP;RoHS;USP
    Series
    Similar to SC0065-AI
    Tubes per pack
    12
  • Pump Tubing, 3-Stop, Puri-Clear™ LL, 1,65 mm ID; 12/PK
    Pump Tubing, 3-Stop, Puri-Clear™ LL, 1,65 mm ID; 12/PK
    1,65 mm
    Blue/Blue/Blue
    406 mm
    Puri-Clear™ LL
    12
    1 * 12 KS
    1 * 12 KS - 1 460,00 Kč
    1 460,00 Kč
    1 * 12 KS
       1 460,00 Kč
     : MFLA95625-38
    Specifications
    Model
    95625-38
    Environmentally Preferable
    Regulatory approvals
    ADCF;Non-DEHP;RoHS;USP
    Series
    Similar to SC0066-AI
    Tubes per pack
    12
  • Pump Tubing, 3-Stop, Puri-Clear™ LL, 1,85 mm ID; 12/PK
    Pump Tubing, 3-Stop, Puri-Clear™ LL, 1,85 mm ID; 12/PK
    1,85 mm
    Green/Green/Green
    406 mm
    Puri-Clear™ LL
    12
    1 * 12 KS
    1 * 12 KS - 1 460,00 Kč
    1 460,00 Kč
    1 * 12 KS
       1 460,00 Kč
     : MFLA95625-40
    Specifications
    Model
    95625-40
    Environmentally Preferable
    Regulatory approvals
    ADCF;Non-DEHP;RoHS;USP
    Series
    Similar to SC0068-AI
    Tubes per pack
    12
  • Pump Tubing, 3-Stop, Puri-Clear™ LL, 2,06 mm ID; 12/PK
    Pump Tubing, 3-Stop, Puri-Clear™ LL, 2,06 mm ID; 12/PK
    2,06 mm
    Purple/purple/purple
    406 mm
    Puri-Clear™ LL
    12
    1 * 12 KS
    1 * 12 KS - 1 460,00 Kč
    1 460,00 Kč
    1 * 12 KS
       1 460,00 Kč
     : MFLA95625-42
    Specifications
    Model
    95625-42
    Environmentally Preferable
    Regulatory approvals
    ADCF;Non-DEHP;RoHS;USP
    Series
    Similar to SC0069-AI
    Tubes per pack
    12
  • Pump Tubing, 3-Stop, Puri-Clear™ LL, 2,29 mm ID; 12/PK
    Pump Tubing, 3-Stop, Puri-Clear™ LL, 2,29 mm ID; 12/PK
    2,29 mm
    Purple/black/Purple
    406 mm
    Puri-Clear™ LL
    12
    1 * 12 KS
    1 * 12 KS - 1 460,00 Kč
    1 460,00 Kč
    1 * 12 KS
       1 460,00 Kč
     : MFLA95625-44
    Specifications
    Model
    95625-44
    Environmentally Preferable
    Regulatory approvals
    ADCF;Non-DEHP;RoHS;USP
    Series
    Similar to SC0070-AI
    Tubes per pack
    12
  • Pump Tubing, 3-Stop, Puri-Clear™ LL, 2,54 mm ID; 12/PK
    Pump Tubing, 3-Stop, Puri-Clear™ LL, 2,54 mm ID; 12/PK
    2,54 mm
    Purple/orange/purple
    406 mm
    Puri-Clear™ LL
    12
    1 * 12 KS
    1 * 12 KS - 1 460,00 Kč
    1 460,00 Kč
    1 * 12 KS
       1 460,00 Kč
     : MFLA95625-46
    Specifications
    Model
    95625-46
    Environmentally Preferable
    Regulatory approvals
    ADCF;Non-DEHP;RoHS;USP
    Series
    Similar to SC0071-AI
    Tubes per pack
    12
  • Pump Tubing, 3-Stop, Puri-Clear™ LL, 2,79 mm ID; 12/PK
    Pump Tubing, 3-Stop, Puri-Clear™ LL, 2,79 mm ID; 12/PK
    2,79 mm
    Purple/white/purple
    406 mm
    Puri-Clear™ LL
    12
    1 * 12 KS
    1 * 12 KS - 1 460,00 Kč
    1 460,00 Kč
    1 * 12 KS
       1 460,00 Kč
     : MFLA95625-48
    Specifications
    Model
    95625-48
    Environmentally Preferable
    Regulatory approvals
    ADCF;Non-DEHP;RoHS;USP
    Series
    Tubes per pack
    12
  • Pump Tubing, 3-Stop, Puri-Clear™ LL, 1,75 mm ID; 12/PK
    Pump Tubing, 3-Stop, Puri-Clear™ LL, 1,75 mm ID; 12/PK
    1,75 mm
    Blue/Green/Blue
    406 mm
    Puri-Clear™ LL
    12
    1 * 12 KS
    1 * 12 KS - 1 170,00 Kč
    1 170,00 Kč
    1 * 12 KS
       1 170,00 Kč
     : MFLA95699-39
    Specifications
    Model
    95699-39
    Environmentally Preferable
    Regulatory approvals
    ADCF;Non-DEHP;RoHS;USP
    Series
    Tubes per pack
    12