Masterflex® Adapter Fittings, Female Luer to Threaded, Straight, Avantor®
Supplier: Avantor Fluid Handling
Certificates
About this item
Easily transition between luer fittings and UNF or NPT threads.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are female thread.
- Environmentally Preferable:
Specifications
- Model:
- 45508-76
- 45508-66
- 45508-60
- 45501-66
- 30800-00
- 45501-60
- Connection type (port 1):
- Female luer
- Female luer
- Female luer
- Female luer
- Female luer
- Female luer
- Connection type (port 2):
- 1/4-28 UNF bottom sealing thread
- 1/4-28 UNF thread
- 10-32 UNF thread
- 1/8-27-NPT thread
- No. of ports:
- 2
- 2
- 2
- 2
- 2
- 2
- Warranty:
- 3 Months
- 3 Months
- 3 Months
Specifications
Product Information
- Catalog #Connection TypeFitting materialFlow patternThread size NPT(M) (port 1)Thread size UNF(M) (port 1)PkAvailabilityPrice
- Fitting, Polypropylene, Straight, Female Luer x 1/8-27 NPT; 25/PKFitting, Polypropylene, Straight, Female Luer x 1/8-27 NPT; 25/PKFemale Luer to ThreadPolypropyleneStraight1/8"251 * 25 KS1 310,00 KčLocal Article Number: MFLA30800-00Specifications:CE certificate
More
Specifications
Model30800-00Environmentally PreferableConnection type (port 1)Female luerConnection type (port 2)1/8-27-NPT threadNo. of ports2Warranty3 MonthsCE certificate - Fitting, Polycarbonate, Straight, Female Luer to Thread Adapter, 10-32 UNF; 25/PKFitting, Polycarbonate, Straight, Female Luer to Thread Adapter, 10-32 UNF; 25/PKFemale Luer to ThreadPolycarbonateStraight10-32251 * 25 KS908,00 KčLocal Article Number: MFLA45501-60Specifications:CE certificateAvantor exclusive
More
Specifications
Model45501-60Environmentally PreferableConnection type (port 1)Female luerConnection type (port 2)No. of ports2Warranty3 Months - Fitting, Polycarbonate, Straight, Female Luer to Thread Adapter, 1/4-28 UNF; 25/PkFitting, Polycarbonate, Straight, Female Luer to Thread Adapter, 1/4-28 UNF; 25/PkFemale Luer to ThreadPolycarbonateStraight1/4"-28251 * 25 KS975,00 KčLocal Article Number: MFLA45501-66Specifications:CE certificateAvantor exclusive
More
Specifications
Model45501-66Environmentally PreferableConnection type (port 1)Female luerConnection type (port 2)No. of ports2Warranty3 Months - Fitting, Polypropylene, Straight, Female Luer to Thread Adapter, 10-32 UNF; 25/PKFitting, Polypropylene, Straight, Female Luer to Thread Adapter, 10-32 UNF; 25/PKFemale Luer to ThreadPolypropyleneStraight10-32251 * 25 KS1 180,00 KčLocal Article Number: MFLA45508-60Specifications:CE certificate
More
Specifications
Model45508-60Environmentally PreferableConnection type (port 1)Female luerConnection type (port 2)10-32 UNF threadNo. of ports2WarrantyCE certificate - Fitting, Polypropylene, Straight, Female Luer to Thread Adapter, with 5/16" Hex to 1/4-28 UNF; 25/PKFitting, Polypropylene, Straight, Female Luer to Thread Adapter, with 5/16" Hex to 1/4-28 UNF; 25/PKFemale Luer to ThreadPolypropyleneStraight1/4"-28251 * 25 KS1 180,00 KčLocal Article Number: MFLA45508-66Specifications:CE certificate
More
Specifications
Model45508-66Environmentally PreferableConnection type (port 1)Female luerConnection type (port 2)1/4-28 UNF threadNo. of ports2WarrantyCE certificate - Fitting, Polypropylene, Straight, Female Luer to Bottom Sealing Thread, 1/4-28 UNF; 25/PKFitting, Polypropylene, Straight, Female Luer to Bottom Sealing Thread, 1/4-28 UNF; 25/PKFemale Luer to ThreadPolypropyleneStraight1/4"-28251 * 25 KS828,00 KčLocal Article Number: MFLA45508-76Specifications:CE certificate
More
Specifications
Model45508-76Environmentally PreferableConnection type (port 1)Female luerConnection type (port 2)1/4-28 UNF bottom sealing threadNo. of ports2WarrantyCE certificate