Tisk…
Surface protection film, Scotchgard™ 2200
Dodavatel: 3M
Scotchgard™
MMMABOFO060EA 4720 CZK
MMMABOFO060
Surface protection film, Scotchgard™ 2200
Ochrana povrchu Papíry ochranné na pracovní desky
Scotchgard™ Surface Protection Film 2200 is a protective film designed to protect stone, vinyl flooring, bathroom surfaces, and walls from everyday challenges. It can be applied to waxed vinyl, sealed concrete, marble, ceramic tile, terrazzo, and more.
This thin protection film is designed to protect surfaces from the everyday challenges they’re subjected to. No special tools or techniques are required to install or remove, and it can be easily removed prior to floor stripping or refinishing. It’s compatible with standard floor cleaning procedures, disinfectants, cleaners, and coatings, helping to reduce the number of stripping and recoating events on floors.