Tisk…

Sulpho-MBS (3-Maleimidobenzoyl-N-hydroxysulphosuccinimide ester sodium salt), Pierce™

Dodavatel: Thermo Fisher Scientific

Synonyms: Sulfo MBS

PIER22312EA 6290 CZK
PIER22312
Sulpho-MBS (3-Maleimidobenzoyl-N-hydroxysulphosuccinimide ester sodium salt), Pierce™
Sulpho-MBS (3-Maleimidobenzoyl-N-hydroxysulphosuccinimide ester sodium salt)

Thermo Scientific Pierce Sulfo-MBS is a water-soluble amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a short aromatic spacer arm (7.3 angstroms).


  • Reactive groups: sulfo-NHS ester and maleimide
  • Reactive towards: amino and sulfhydryl groups
  • Non-cleavable
  • Water-soluble (compare to MBS)
  • Membrane impermeable, allowing cell surface labeling


Upozornení: For Research Use Only. Not for use in diagnostic procedures.

Vzorec: C₁₅H₉N₂O₉S·Na
MW: 416,295 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01318594
CAS číslo: 92921-25-0

Objednat

Výsledky specifikačního testu

Product Line Pierce™
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Maleimide, Sulfo-NHS Ester
Spacer Short
Form Powder
Quantity 50 mg
Type Crosslinker
Solubility Water

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR