Tisk…

Sulpho-MBS (3-Maleimidobenzoyl-N-hydroxysulphosuccinimide ester sodium salt)

Dodavatel: G Bioscienes
Varování

Synonyms: Sulfo MBS

BC12EA 16980 CZK
BC12
Sulpho-MBS (3-Maleimidobenzoyl-N-hydroxysulphosuccinimide ester sodium salt)
Sulpho-MBS (3-Maleimidobenzoyl-N-hydroxysulphosuccinimide ester sodium salt)

m-Maleimidobenzoyl-N-hydroxysulfosuccinimide esterHeterobifunctional Cross Linker


Upozornení: For Research Use Only. Not for use in diagnostic procedures.

Vzorec: C₁₅H₉N₂O₉S·Na
MW: 416,295 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01318594
CAS číslo: 92921-25-0

Objednat

Výsledky specifikačního testu

Spacer Arm (Å) 9.9
Reactive Toward Primary Amine + Sulfhydryl
Membrane Permeable NO
Water Soluble YES
Cleavable/ Reversible NO

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR