Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Czech Republic
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0,00 Kč
Order Entry
Home
Standards
Organic Standards
Estradiol Valerate, Reference Standard
Estradiol Valerate, Reference Standard
Estradiol Valerate, Reference Standard
1254009.
:
USP
:
Estradiol Valerate, Reference Standard
1254009.
:
USP
:
: 1254009
: USPH1254009
10 980,00 Kč
1 * 100 mg
-
+
Pack type:
Vial
Pk:
100 mg
Standards:
USP reference standard
CAS No.:
979-32-8
Description:
Estradiol Valerate
Seq:
04
Formula:
C₂₃H₃₂O₃
Description:
Estradiol Valerate