Fexofenadine Related Compounds, Reference Standard
: USP
Specifications
- Synonym(s):(+/-)-4-[1-hydroxy-4-[4-(hydroxydiphenylmethyl)-1-piperidinyl]-butyl]-isopropylbenzene hydrochloride
- Description:Fexofenadine Related Compound C
- CAS No.:76811-98-8
- Formula:C₃₂H₃₇NO₄
Specifications
- StandardsPack typePk
- Fexofenadine Related Compound AFexofenadine Related Compound A1270388.1270388USP reference standardVial25 mg1 * 25 mg35 060,00 Kč: 1270388: USPH1270388
- Fexofenadine Related Compound BFexofenadine Related Compound B1270399.1270399USP reference standardVial25 mg1 * 25 mg38 480,00 Kč: 1270399: USPH1270399
- Fexofenadine Related Compound CFexofenadine Related Compound C1270446.1270446USP reference standardVial25 mg1 * 25 mg36 770,00 Kč: 1270446: USPH1270446